missing translation for 'onlineSavingsMsg'
Learn More

GPANK1/BAT4 Antibody, Novus Biologicals™

Product Code. 18326898 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326898 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326898 Supplier Novus Biologicals Supplier No. H00007918B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

GPANK1/BAT4 Polyclonal antibody specifically detects GPANK1/BAT4 in Human samples. It is validated for Western Blot, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPANK1/BAT4
Applications Western Blot, Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Dot Blot
Formulation PBS (pH 7.4)
Gene Accession No. NP_149417
Gene Alias 6S54E, ANKRD59Protein G5, Ankyrin repeat domain-containing protein 59, BAT4G patch domain containing 10, D6S54EG patch domain and ankyrin repeats-containing protein 1, G patch domain and ankyrin repeats 1, G patch domain-containing protein 10, G5HLA-B-associated transcript 4, GPATCH10HLA-B associated transcript 4, protein BAT4
Host Species Mouse
Immunogen BAT4 (NP_149417, 1 a.a. - 356 a.a.) full-length human protein. MSRPLLITFTPATDPSDLWKDGQQQPQPEKPESTLDGAAARAFYEALIGDESSAPDSQRSQTEPARERKRKKRRIMKAPAAEAVAEGASGRHGQGRSLEAEDKMTHRILRAAQEGDLPELRRLLEPHEAGGAGGNINARDAFWWTPLMCAARAGQGAAVSYLLGRGAAWVGVCELSGRDAAQLAEEAGFPEVARMVRESHGETRSPENRSPTPSLQYCENCDTHFQDSNHRTSTAHLLSLSQGPQPPNLPLGVPISSPGFKLLLRGGWEPGMGLGPRGEGRANPIPTVLKRDQEGLGYRSAPQPRVTHFPAWDTRAVAGRERPPRVATLSWREERRREEKDRAWERDLRTYMNLEF
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7918
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.