missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPAM Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£153.00
Specifications
| Antigen | GPAM |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
GPAM Polyclonal specifically detects GPAM in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| GPAM | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Rabbit | |
| Human | |
| NP_065969 | |
| 57678 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of the human GPAT1. The immunogen is located within amino acids 730 - 780 of GPAT1. Amino Acid Squence: SGPVPEPEYLQKLHK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 92 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| EC 2.3.1, EC 2.3.1.15, glycerol-3-phosphate acyltransferase 1, mitochondrial, glycerol-3-phosphate acyltransferase, mitochondrial, GPAT, GPAT-1, GPAT1mitochondrial, MGC26846 | |
| GPAM | |
| IgG | |
| Affinity Purified | |
| At least four isoforms of GPAT1 are known to exist; this antibody will detect only the two largest isoforms. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title