missing translation for 'onlineSavingsMsg'
Learn More

GPAM Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18419822 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419822 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419822 Supplier Novus Biologicals Supplier No. NBP176907AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPAM Polyclonal antibody specifically detects GPAM in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPAM
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias EC 2.3.1, EC 2.3.1.15, glycerol-3-phosphate acyltransferase 1, mitochondrial, glycerol-3-phosphate acyltransferase, mitochondrial, GPAT, GPAT-1, GPAT1mitochondrial, MGC26846
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of the human GPAT1. The immunogen is located within amino acids 730 - 780 of GPAT1. Amino Acid Squence: SGPVPEPEYLQKLHK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 57678
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.