missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPALPP1 Polyclonal Antibody

GPALPP1 Polyclonal Antibody
Brand: Thermo Scientific PA558613
This item is not returnable.
View return policy
Description
GPALPP1 Polyclonal Antibody for IHC
Specifications
| GPALPP1 | |
| Polyclonal | |
| Unconjugated | |
| GPALPP1 | |
| 1200011I18Rik; AD029; bA245H20.2; C6orf72; dJ12G14.2; GINM1; glycoprotein integral membrane 1; glycoprotein integral membrane protein 1; GPALPP motifs containing 1; GPALPP motifs-containing protein 1; Gpalpp1; hypothetical protein LOC532698; hypothetical protein LOC55425; Kiaa1704; lipopolysaccharide specific response-7 protein; lipopolysaccharide-specific response protein 7; LSR7; RGD1307254; UNQ710/PRO1361 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55425 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IXQ4 | |
| GPALPP1 | |
| Recombinant protein corresponding to Human GPALPP1. Recombinant protein control fragment (Product #RP-97945). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction