Learn More
Invitrogen™ GPAA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589485
Description
Immunogen sequence: AWLERTMRSV GLEVYTQSFS RKLPFPDETH ERYMVSGTNV YGILRAPRAA STESLVLTVP CGSDSTNSQA VGLLLALAAH FRGQIYWAKD IVFLVTEHDL LGTEAWLEAY HDVNVTGMQS SPLQGRAGAI QAAVALELSS DVVTSLDVAV EGLNGQLPNL DLLNLFQTFC QKGGLLCTLQ GKLQPEDWTS LDGPLQGLQT LLLMVLRQAS GRPHGSHGLF L; Positive Samples: Mouse heart; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.
Posttranslational glycosylphosphatidylinositol (GPI) anchor attachment serves as a general mechanism for linking proteins to the cell surface membrane. The protein encoded by this gene presumably functions in GPI anchoring at the GPI transfer step. The mRNA transcript is ubiquitously expressed in both fetal and adult tissues. The anchor attachment protein 1 contains an N-terminal signal sequence, 1 cAMP- and cGMP-dependent protein kinase phosphorylation site, 1 leucine zipper pattern, 2 potential N-glycosylation sites, and 8 putative transmembrane domains.
Specifications
| GPAA1 | |
| Polyclonal | |
| Unconjugated | |
| GPAA1 | |
| anchor attachment protein 1; anchor attachment protein 1 (Gaa1p, yeast) homolog; C80044; GAA1; GAA1 protein homolog; glycophosphatidylinositol anchor attachment 1; glycosylphosphatidylinositol anchor attachment 1; glycosylphosphatidylinositol anchor attachment 1 (GPAA1); glycosylphosphatidylinositol anchor attachment 1 protein; glycosylphosphatidylinositol anchor attachment protein 1 homolog; Gpaa1; GPAA1 protein; GPAA1P anchor attachment protein 1 homolog; GPI anchor attachment protein 1; GPI transamidase subunit; hGAA1; mGAA1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14731 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.88 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9WTK3 | |
| GPAA1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 90-310 of human GPAA1 (NP_0037921). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.