missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144819
This item is not returnable.
View return policy
Description
Immunogen sequence: ADPEIKRGSG WRPTGCSQPR VMFMTAEPQA RSYPREGSWH GRRLKDWRVW SVEAGGQRLQ LWKRGHAASS WCSIREPFGQ CLSVCLP Antigen sequence identity: Mouse 25%; Rat 22%.
Glycoprotein VI (GP6) is a 58-kD platelet membrane glycoprotein that plays a crucial role in the collagen-induced activation and aggregation of platelets. Upon injury to the vessel wall and subsequent damage to the endothelial lining, exposure of the subendothelial matrix to blood flow results in deposition of platelets. Collagen fibers are the most thrombogenic macromolecular components of the extracellular matrix, with collagen types I, III, and VI being the major forms found in blood vessels. Platelet interaction with collagen occurs as a 2-step procedure: (1) the initial adhesion to collagen is followed by (2) an activation step leading to platelet secretion, recruitment of additional platelets, and aggregation. In physiologic conditions, the resulting platelet plug is the initial hemostatic event limiting blood loss. However, exposure of collagen after rupture of atherosclerotic plaques is a major stimulus of thrombus formation associated with myocardial infarction or stroke.
Specifications
| GP6 | |
| Polyclonal | |
| Unconjugated | |
| GP6 | |
| 9830166G18Rik; BDPLT11; Glycoprotein 5; glycoprotein 6; glycoprotein 6 (platelet); glycoprotein VI (platelet); glycoprotein VI platelet; Gm469; GP6; GPIV; GPVI; platelet collagen receptor; platelet glycoprotein VI | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51206 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HCN6 | |
| GP6 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 318-404. Recombinant protein control fragment (Product #RP-110189). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction