Learn More
Invitrogen™ gp210 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143938
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human MCF-7 whole cell, human HEK293 whole cell, rat thymus tissue. IHC: human liver tissue, human liver tissue, human appendiceal adenocarcinoma tissue. ICC/IF: MCF-7 cell. Flow: JK cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
NUP210 nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex.
Specifications
| gp210 | |
| Polyclonal | |
| Unconjugated | |
| NUP210 | |
| 9830001L10; AI836801; FLJ22389; gp190; GP210; integral membrane nucleoporin gp210; Kiaa0906; Nuclear envelope pore membrane protein POM 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nuclear pore protein gp210; nucleoporin 210; nucleoporin 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nucleoporin 210kDa; nucleoporin 210kDa L homeolog; nucleoporin Nup210; NUP210; nup210.L; POM210; pore membrane protein of 210 kDa; PSEC0245; XELAEV_18001839mg | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23225, 58958 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P11654, Q8TEM1 | |
| NUP210 | |
| E.coli-derived human GP210/NUP210 recombinant protein (Position: R221-K1724). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.