missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ gp210 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110051
This item is not returnable.
View return policy
Description
Immunogen sequence: QNVTAEDTDS IGLALFAPHS SRNYQQHWIL VTCQALGEQV IALSVGNKPS LTNPFPAVEP AVVKFVCAPP SRLTLAPVYT SPQLDMSCPL LQQNKQVVPV SSHRNPRLDL AAYDQEGRRF DNFSSLSIQW ESTRPVLASI EPELPMQLVS QDDESGQKKL HGLQAILVHE ASGTTAITAT ATGYQESHLS SARTKQPHDP L.
NUP210 nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex.
Specifications
| gp210 | |
| Polyclonal | |
| Unconjugated | |
| NUP210 | |
| 9830001L10; AI836801; FLJ22389; gp190; GP210; integral membrane nucleoporin gp210; Kiaa0906; Nuclear envelope pore membrane protein POM 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nuclear pore protein gp210; nucleoporin 210; nucleoporin 210; nuclear pore membrane glycoprotein 210; nuclear pore membrane protein 210; nucleoporin 210kDa; nucleoporin 210kDa L homeolog; nucleoporin Nup210; NUP210; nup210.L; POM210; pore membrane protein of 210 kDa; PSEC0245; XELAEV_18001839mg | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23225, 54563, 58958 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.87 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P11654, Q8TEM1, Q9QY81 | |
| NUP210 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 680-880 of human NUP210 (NP_079199.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction