missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553375
This item is not returnable.
View return policy
Description
Immunogen sequence: TVEDKCEKAC RPEEECLALN STWGCFCRQD LNSSDVHSLQ PQLDCGPREI KVKVDKCLLG GLGLGEEVIA YLRDPNCSSI LQTEERNWVS VTSPVQASAC RNILERNQTH AIYK Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 64%.
GP2 is component of pancreatic secretory (zymogen) granule. The exact function of GP2 remains unknown.
Specifications
| GP2 | |
| Polyclonal | |
| Unconjugated | |
| GP2 | |
| 2310037I18Rik; AV060639; glycoprotein 2; glycoprotein 2 (zymogen granule membrane); glycoprotein 80; GP2; gp80; Pancreatic secretory granule membrane major glycoprotein GP2; pancreatic zymogen granule membrane associated protein GP2; pancreatic zymogen granule membrane protein GP-2; secretory (zymogen) granule membrane glycoprotein GP2; ZAP75; zymogen granule membrane glycoprotein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2813 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P55259 | |
| GP2 | |
| Recombinant protein corresponding to Human GP2. Recombinant protein control fragment (Product #RP-90453). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction