missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP2 (Glycoprotein 2) / ZAP75 Mouse, Clone: GP2/1803, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2813-MSM3-P0
Additional Details : Weight : 0.01000kg
Specifications
GP2 (Glycoprotein 2) / ZAP75 | |
Cell Surface and Secreted | |
GP2/1803 | |
Unconjugated | |
Mouse | |
59kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
2813 | |
IgG2a κ |
ELISA, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human GP2 protein (apprx. aa 35-179) | |
PANC-1 Cells. Pancreas | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |