missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP2 Antibody (GP2/3134R), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08243H
This item is not returnable.
View return policy
Description
GP2 Monoclonal antibody specifically detects GP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifications
| GP2 | |
| Monoclonal | |
| HRP | |
| DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75 | |
| Recombinant fragment of human GP2 protein (around aa 35-179) (exact sequence is proprietary) (Uniprot: P55259) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| GP2/3134R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 2813 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction