missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP1BB Polyclonal Antibody

GP1BB Polyclonal Antibody
Brand: Thermo Scientific PA5104021
This item is not returnable.
View return policy
Description
GP1BB Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| GP1BB | |
| Polyclonal | |
| Unconjugated | |
| GP1BB | |
| Antigen CD42b-beta; BDPLT1; BS; CD42c; glycoprotein Ib (platelet), beta polypeptide; glycoprotein Ib platelet beta subunit; glycoprotein Ib, beta polypeptide; Gp1bb; GP-Ib beta; GPIbB; GPIbbeta; GPIb-beta; nuclear localization signal deleted in velocardiofacial syndrome; platelet glycoprotein Ib beta chain; platelet membrane glycoprotein Ib beta; truncated platelet membrane glycoprotein Ib beta | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14724, 2812 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P13224, P56400 | |
| GP1BB | |
| A synthesized peptide derived from human GP1BB(Accession P13224), corresponding to amino acid residues A84-E134. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction