missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ gp120 (AC10.0) (HIV-1/Clade B) (AY835446, 36 a.a. - 514 a.a.) Partial Recombinant Protein with His tag expressed in 293 cells
Used for SDS-PAGE
Brand: Abnova™ P5096.50ug
Additional Details : Weight : 0.02000kg
Specifications
AY835446 | |
SDS-PAGE | |
gp120 (AC10.0) (HIV-1/Clade B) Recombinant Protein | |
SDS-PAGE Result | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
GP120 | |
Recombinant | |
Mammalian cell (293) expression system | |
≥95% by SDS-PAGE |
1 ug/uL | |
Liquid | |
Mammalian cell (293) expression system | |
50 ug | |
RUO | |
Human | |
His | |
Liquid |