missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gp100 / Melanosome / PMEL17 / SILV Mouse, Clone: HMB45 + PMEL/783, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6490-MSM4-P1
This item is not returnable.
View return policy
Specifications
| gp100 / Melanosome / PMEL17 / SILV | |
| Cytoplasmic | |
| HMB45 + PMEL/783 | |
| Unconjugated | |
| NP_001307051 | |
| Mouse | |
| 90-100kDa | |
| Protein A/G Purified | |
| RUO | |
| 6490 | |
| IgG |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| SILV | |
| Extract of pigmented melanoma metastases from lymph nodes (HMB45); Recombinant human SILV protein (PMEL/783) | |
| SK-MEL-28 cells or Melanoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction