missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP-IX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | GP-IX |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18101549
|
Novus Biologicals
NBP2-38018 |
0.1 mL |
£488.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675225
|
Novus Biologicals
NBP2-38018-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GP-IX Polyclonal specifically detects GP-IX in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GP-IX | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CD42a, CD42a antigen, Glycoprotein 9, glycoprotein IX (platelet), GP-IX, GPIXplatelet glycoprotein IX | |
| GP9 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P14770 | |
| 2815 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: HGLTALPALPARTRHLLLANNSLQSVPPGAFDHLPQLQTLDVTQNPWHCDC | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title