missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOT2 Antibody (8B11), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59428PE
This item is not returnable.
View return policy
Description
GOT2 Monoclonal antibody specifically detects GOT2 in Human, Rat samples. It is validated for Flow Cytometry
Specifications
| GOT2 | |
| Monoclonal | |
| PE | |
| aspartate aminotransferase, mitochondrial, EC 2.6.1, EC 2.6.1.1, FABP-1, FABPpm, Fatty acid-binding protein, FLJ40994, Glutamate oxaloacetate transaminase 2, glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase2), KAT4, KATIV, kynurenine aminotransferase IV, mAspAT, mitAAT, Plasma membrane-associated fatty acid-binding protein, Transaminase A | |
| Recombinant human GOT2 (30-430aa) purified from E.coli. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 2806 | |
| Store at 4°C in the dark. | |
| IgG3 κ |
| Flow Cytometry | |
| 8B11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction