missing translation for 'onlineSavingsMsg'
Learn More

GOT2 Antibody (8B11), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18428882 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428882 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428882 Supplier Novus Biologicals Supplier No. NBP259428MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GOT2 Monoclonal antibody specifically detects GOT2 in Human, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOT2
Applications Flow Cytometry
Classification Monoclonal
Clone 8B11
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias aspartate aminotransferase, mitochondrial, EC 2.6.1, EC 2.6.1.1, FABP-1, FABPpm, Fatty acid-binding protein, FLJ40994, Glutamate oxaloacetate transaminase 2, glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase2), KAT4, KATIV, kynurenine aminotransferase IV, mAspAT, mitAAT, Plasma membrane-associated fatty acid-binding protein, Transaminase A
Host Species Mouse
Immunogen Recombinant human GOT2 (30-430aa) purified from E.coli.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2806
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.