missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOT2 Antibody (3E9), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£161.00
Specifications
| Antigen | GOT2 |
|---|---|
| Clone | 3000000000 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
GOT2 Monoclonal specifically detects GOT2 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| GOT2 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with 0.03% Sodium Azide | |
| aspartate aminotransferase, mitochondrial, EC 2.6.1, EC 2.6.1.1, FABP-1, FABPpm, Fatty acid-binding protein, FLJ40994, Glutamate oxaloacetate transaminase 2, glutamic-oxaloacetic transaminase 2, mitochondrial (aspartate aminotransferase2), KAT4, KATIV, kynurenine aminotransferase IV, mAspAT, mitAAT, Plasma membrane-associated fatty acid-binding protein, Transaminase A | |
| GOT2 | |
| IgG1 | |
| Unpurified | |
| 47 kDa |
| 3000000000 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P00505 | |
| 2806 | |
| Purified recombinant fragment of human GOT2 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title