missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GOT1 Recombinant Rabbit Monoclonal Antibody (23GB5585), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553904
This item is not returnable.
View return policy
Description
GOT1 is a glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.
Specifications
| GOT1 | |
| Recombinant Monoclonal | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05201, P13221, P17174 | |
| GOT1 | |
| A synthesized peptide derived from human Aspartate Aminotransferase (150-230AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5585 | |
| Unconjugated | |
| GOT1 | |
| AI789014; aspartate aminotransferase 1; aspartate aminotransferase, cytoplasmic; aspartate transaminase 1; Aspat; AST1; ASTQTL1; cAspAT; cCAT; Cysteine aminotransferase, cytoplasmic; cysteine transaminase, cytoplasmic; cytosolic aspartate aminotransferase; Gaspat; GIG18; Glutamate oxaloacetate transaminase 1; glutamate oxaloacetate transaminase 1, soluble; glutamic-oxaloacetic transaminase 1; glutamic-oxaloacetic transaminase 1, soluble; glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1); GOT1; Got-1; growth-inhibiting protein 18; testis secretory sperm-binding protein Li 196a; transaminase A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14718, 24401, 2805 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction