missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14886-1-AP-150UL
This item is not returnable.
View return policy
Description
GOT1 is a glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.Specifications
| GOT1 | |
| Polyclonal | |
| Unconjugated | |
| GOT1 | |
| AATC, CAT, GIG18, GOT1, Transaminase A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14718, 24401, 2805 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05201, P13221, P17174 | |
| GOT1 | |
| GOT1 Fusion Protein Ag6671 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction