missing translation for 'onlineSavingsMsg'
Learn More

Gossypol, R&D Systems™

Product Code. 18702794
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18702794 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18702794 Supplier Tocris Bioscience Supplier No. 1964/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Proapoptotic; downregulates Bcl-2 and Bcl-XL

Lipophilic agent derived from cottonseed. Exhibits multiple biological effects including antifertility and anticancer activity. Proapoptotic; downregulates Bcl-2 and Bcl-XL.

R-enantiomer also available.

TRUSTED_SUSTAINABILITY

Specifications

CAS 303-45-7
Molecular Formula C30H30O8
MDL Number MFCD00017352
Quantity 50 mg
Synonym gossypol, --gossypol, +-gossypol, pogosin, tash 1, racemic-gossypol, r---gossypol, r-gossypol, +--gossypol, +/--gossypol
InChI Key QBKSWRVVCFFDOT-UHFFFAOYSA-N
SMILES CC(C)C1=C(O)C(O)=C(C=O)C2=C(O)C(=C(C)C=C12)C1=C(O)C2=C(C=O)C(O)=C(O)C(C(C)C)=C2C=C1C
IUPAC Name 1,1',6,6',7,7'-hexahydroxy-3,3'-dimethyl-5,5'-bis(propan-2-yl)-[2,2'-binaphthalene]-8,8'-dicarbaldehyde
Molecular Weight (g/mol) 518.56
PubChem CID 3503
ChEBI CHEBI:28584
Formula Weight 518.56
Percent Purity >98%
Product Line Tocris
Chemical Name or Material Gossypol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.