missing translation for 'onlineSavingsMsg'
Learn More

Gossypol Acetic Acid, TRC

Product Code. 30136638
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136638 2.5 g 2.5g
30040729 100 mg 100mg
30011990 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136638 Supplier Toronto Research Chemicals Supplier No. TRCG7679002.5G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12542-36-8
Molecular Formula C30 H30 O8 . C2 H4 O2
Molecular Weight (g/mol) 578.61
Synonym Gossypol acetic acid, 1,1',6,6',7,7'-Hexahydroxy-3,3'-dimethyl-5,5'-bis(1-methylethyl)-[2,2'-binaphthalene]-8,8'-dicarboxaldehyde Acetic Acid, 1,1',6,6',7,7'-Hexahydroxy-5,5'-diisopropyl-3,3'-dimethyl-[2,2'-binaphthalene]-8,8'-dicarboxaldehyde Acetic Acid, (±)-Gossypol Acetic Acid, 1,1',6,6',7,7'-Hexahydroxy-3,3'-dimethyl-5,5'-diisopropyl-2,2'-binaphthyl-8,8'-dialdehyde Acetic Acid, 1,1',6,6',7,7'-Hexahydroxy-3,3'-dimethyl-5,5'-diisopropyl[2,2'-binaphthalene]-8,8'-dicarboxaldehyde Acetic Acid, 1,1',6,6',7,7'-Hexahydroxy-5,5'-diisopropyl-3,3'-dimethyl-2,2'-binaphthalene-8,8'-dicarboxaldehyde Acetic Acid, 1,6,7,1',6',7'-Hexahydroxy-5,5'-diisopropyl-3,3'-dimethyl-[2,2']binaphthalenyl-8,8'-dicarboxaldehyde Acetic Acid, 2,2'-Bis[1,6,7-trihydroxy-3-methyl-5-isopropyl-8-aldehydonaphthyl] Acetic Acid, 2,2'-Bis[8-formyl-1,6,7-trihydroxy-5-isopropyl-3-methylnaphthyl] Acetic Acid, BL 193 Acetic Acid, Gossypol Acetic Acid, NSC 56817 Acetic Acid, NSC 624336 Acetic Acid, No Fertil Acetic Acid, Pogosin Acetic Acid, Tash 1 Acetic Acid
IUPAC Name acetic acid;7-(8-formyl-1,6,7-trihydroxy-5-isopropyl-3-methyl-2-naphthyl)-2,3,8-trihydroxy-4-isopropyl-6-methyl-naphthalene-1-carbaldehyde
SMILES CC(C)c1c(O)c(O)c(C=O)c2c(O)c(c(C)cc12)c3c(C)cc4c(C(C)C)c(O)c(O)c(C=O)c4c3O.CC(=O)O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C30H30O8.C2H4O2/c1-11(2)19-15-7-13(5)21(27(35)23(15)17(9-31)25(33)29(19)37)22-14(6)8-16-20(12(3)4)30(38)26(34)18(10-32)24(16)28(22)36;1-2(3)4/h7-12,33-38H,1-6H3;1H3,(H,3,4)
Formula Weight 578.2152
Chemical Name or Material Gossypol Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.