missing translation for 'onlineSavingsMsg'
Learn More

Gossypol (acetic acid), MedChemExpress

Product Code. 16451876
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451876 500 mg 500mg
16401276 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451876 Supplier Medchem Express Supplier No. HY17510

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Gossypol acetic acid ((±)-Gossypol-acetic acid) binds to Bcl-xL protein and Bcl-2 protein with Kis of 0.5-0.6 μM and 0.2-0.3 mM, respectively.

Gossypol acetic acid ((±)-Gossypol-acetic acid) binds to Bcl-xL protein and Bcl-2 protein with Kis of 0.5-0.6 μM and 0.2-0.3 mM, respectively.

Specifications

Chemical Name or Material Gossypol (acetic acid)
CAS 12542-36-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C32H34O10
Quantity 500 mg
Synonym (±)-Gossypol-acetic acid BL-193 (acetic acid)
Solubility Information DMSO : 25 mg/mL (43.21 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC1=C(C2=C(O)C(C(C=O)=C(O)C(O)=C3C(C)C)=C3C=C2C)C(C)=CC4=C(C(C)C)C(O)=C(O)C(C=O)=C41.O=C(O)C
Molecular Weight (g/mol) 578.61
Formula Weight 578.61
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.