Learn More
Gosselin™ Straight Container
Product Code. 15488824
Click to view available options
Capacity (Metric):
60 mL
125 mL
180 mL
Closure Color:
Blue
Green
Red
Unit Size:
Pack of 264
Pack of 380
Pack of 600
Pack of 700
Product Code. 15488824
Brand: Gosselin™ TP52C004
Corning™ Gosselin™ Straight Container, Specimen Vials, PP come in different sizes and quantities, and are available sterile and non-sterile.
  • Clear and break-resistant polypropylene (PP)
  • Available in different sizes and quantities
  • Color-coded screw caps
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Sterility Sterile
Capacity (Metric) 125 mL
Closure Color Green
No. per Case 380/Cs.
No. per Pack 380/Pk.