Learn More
Gosselin™ Screw Cap
Shop All Corning Products
Click to view available options
Color:
Red
Unit Size:
Pack of 3150
Description
- Helps to prevent contact with the contents of the tube when screwing and unscrewing
- Helps to prevent micro-projections when opening the tube.
Specifications
Specifications
| Material | Polyethylene |
| Color | Red |
| No. per Pack | 450/Pk. |
| No. per Case | 3150/Cs. |