missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Biohazard Pouch

Double-pocketed, self-adhesive biohazard bags printed with a prominent biohazard logo. Gosselin™ Biohazard Pouch is designed for decontaminating laboratory waste via autoclaving.

Brand:  Gosselin™ SFAB1727-08

Code : A0

Additional Details : Weight : 0.20000kg

Product Code. 11771605

  • £473.82 / Pack of 2000
Estimated Shipment: 19-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Durable polypropylene construction
  • Conical shape
  • Flat bottom
  • Red screw-cap closure
Specifications

Specifications

Self-Adhesive Closure
170 mm
40 μm
270 mm
2000/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers