missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Gosselin™ Polyethylene Octagonal Thiosulfate Sodium Bottles

Catalog Number 15464570
Click to view available options
Material:
Polyethylene
This item is not returnable. View return policy
This item is not returnable. View return policy

Break-resistant polyethylene terephthalate (PET) or High-density Polyethylene (HDPE)

  • Break-resistant polyethylene terephthalate (PET) or High-density Polyethylene (HDPE)
  • Molded graduations
  • Leak-proof tamper-evident HDPE screw caps are available for the 500 mL and 1 L bottle not the 250 mL.
  • Available with or without sodium thiosulfate
  • Traceable labels
  • Large openings minimize risk of contamination
  • Film-packed or individually bagged
TRUSTED_SUSTAINABILITY
Diameter (Metric) Inner Neck 31.7 mm
Material Polyethylene
Length (Metric) 96mm
Width (Metric) 96mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'