missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Autoclavable Bags
Click to view available options
Capacity (Metric):
3 L
9 L
14 L
33 L
Material:
HDPE
Polypropylene
Width (Metric):
25.5 cm
30 cm
40 cm
60 cm
Height (Metric):
40 cm
50 cm
60 cm
78 cm
Unit Size:
Pack of 450
Pack of 500
Pack of 750
Description
- HDPE bags
- Thermal resistance 134°C
- Transparent
Order Info
Unprinted bags are packaged in boxes. Printed bags are packaged in rolls.
Specifications
Specifications
| Capacity (Metric) | 9 L |
| Capacity (English) | 2.37 gal. |
| Color | Natural |
| Height (Metric) | 60 cm |
| Width (Metric) | 40 cm |
| Autoclavable | Autoclavable |
| Includes | Biohazard symbol |
| For Use With (Application) | Clinical, dairy, environment, industries, microbiology |
| Width (English) | 15.74 in. |
| Thickness | 40 μm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction