missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GoSlo SR 5-69

Description
GoSlo SR 5-69 is a BK channel activator (EC50 = 251 nM). Shifts the voltage required for half maximal negatively (DV1/2 = -104 mV).
Specifications
Specifications
| Chemical Name or Material | Sodium 1-Amino-9,10-dioxo-4-((5,6,7,8-tetrahydronaphthalen-2-yl)amino)-9,10-dihydroanthracene-2-sulfonate |
| CAS | 1363419-31-1 |
| Quantity | 10 mg |
| Target | Ca2+-Activated Potassium Channel Activators |
| Molecular Formula | C24H19N2NaO5S |
| Formula Weight | 2576 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?