missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goosecoid Antibody (OTI1D7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47790
This item is not returnable.
View return policy
Description
Goosecoid Monoclonal specifically detects Goosecoid in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Goosecoid | |
| Monoclonal | |
| 0.7 mg/mL | |
| Western Blot 1:500-1000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50 | |
| GSC | |
| Human recombinant protein fragment corresponding to amino acids 107-257 of human GSC (NP_776248) produced in E.coli. | |
| Affinity Purified | |
| RUO | |
| 145258 | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI1D7 | |
| Unconjugated | |
| goosecoid homeobox, homeobox protein goosecoid | |
| Mouse | |
| 28 kDa | |
| 0.1 mL | |
| Primary | |
| This antibody is specific for Homo sapiens goosecoid homeobox (GSC). | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction