missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goosecoid Antibody (OTI1D7), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72409CL1
This item is not returnable.
View return policy
Description
Goosecoid Monoclonal antibody specifically detects Goosecoid in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
| Goosecoid | |
| Monoclonal | |
| CoraFluor 1 | |
| goosecoid homeobox, homeobox protein goosecoid | |
| Human recombinant protein fragment corresponding to amino acids 107-257 of human GSC (NP_776248) produced in E.coli. | |
| 0.1 mL | |
| Stem Cells | |
| 145258 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| OTI1D7 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction