missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GON4L Polyclonal Antibody

Product Code. 15918804
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15918804 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15918804 Supplier Invitrogen™ Supplier No. PA566076

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PMEEQDNEESE KRRKKKKGTK RKRDGRGQEG TLAYDLKLDD MLDRTLEDGA KQHNLTAVNV RNILHEVITN EHVVAMMKAA ISETEDMPMF EPKMTRSKLK EVV Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.

Has transcriptional repressor activity, probably as part of a complex with YY1, SIN3A AND HDAC1. Required for B cell lymphopoiesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GON4L
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GON4L
Gene Accession No. Q3T8J9
Gene Alias 2610100B20Rik; Dingo; Gon4; GON-4; GON-4 homolog; gon-4 like; GON4L; gon-4-like (C. elegans); GON-4-like protein; KIAA1606; Protein GON4; YARP; YY1AP related protein; YY1AP-related protein1
Gene Symbols GON4L
Host Species Rabbit
Immunogen Recombinant Human GON4L. Recombinant protein control fragment (Product #RP-106768).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54856
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.