missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GON4L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568195
This item is not returnable.
View return policy
Description
GON4L Polyclonal Antibody for Western Blot
Has transcriptional repressor activity, probably as part of a complex with YY1, SIN3A AND HDAC1. Required for B cell lymphopoiesis.
Specifications
| GON4L | |
| Polyclonal | |
| Unconjugated | |
| GON4L | |
| 2610100B20Rik; Dingo; Gon4; GON-4; GON-4 homolog; gon-4 like; GON4L; gon-4-like (C. elegans); GON-4-like protein; KIAA1606; Protein GON4; YARP; YY1AP related protein; YY1AP-related protein1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54856 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 30% glycerol, 0.87% NaCl and 0.01% sodium azide; pH 7.3 | |
| Q3T8J9 | |
| GON4L | |
| Synthesized peptide derived from human GON4L. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction