missing translation for 'onlineSavingsMsg'
Learn More

GOLPH4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18344986 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344986 100 μL 100µL
18362054 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18344986 Supplier Novus Biologicals Supplier No. NBP303684100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GOLPH4 Polyclonal specifically detects GOLPH4 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLPH4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias GIMPc, GIMPCcis, golgi integral membrane protein 4, Golgi phosphoprotein 4130 kDa golgi-localized phosphoprotein, Golgi phosphoprotein of 130 kDa, GPP130Golgi-localized phosphoprotein of 130 kDa
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 34-300 of human GOLPH4 (NP_055313.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 27333
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.