missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GOLPH3L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546472
This item is not returnable.
View return policy
Description
Peptide sequence: EISKNSEKKM ESEEDSNWEK SPDNEDSGDS KDIRLTLMEE VLLLGLKDKE Sequence homology: Human: 100%; Mouse: 79%; Rat: 79%.
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is localized at the Golgi stack and may have a regulatory role in Golgi trafficking.
Specifications
| GOLPH3L | |
| Polyclonal | |
| Unconjugated | |
| GOLPH3L | |
| 2010204I15Rik; AU015439; AW549227; C77637; golgi phosphoprotein 3 like; golgi phosphoprotein 3-like; Golph3l; GPP34R; GPP34-related protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55204 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H4A5 | |
| GOLPH3L | |
| Synthetic peptide directed towards the N-terminal of human GOLPH3L (aa 12-61). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction