Learn More
GOLPH3 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-19112-100UL
Description
The GOLPH3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GOLPH3. This antibody recognizes human, mouse, rat antigen. The GOLPH3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| GOLPH3 | |
| 1000 μg/mL | |
| Coat protein GPP34, FLJ90675, Golgi phosphoprotein 3, GOLPH3, GOPP1, GPP34, MIDAS | |
| 100 μL | |
| 64083 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |