missing translation for 'onlineSavingsMsg'
Learn More

GOLPH3 Polyclonal Antibody

Product Code. 12848952
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12848952 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12848952 Supplier Invitrogen Supplier No. PA520830

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GOLPH3 Polyclonal Antibody

GOLPH3 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLPH3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Golph3
Gene Accession No. Q9CRA5, Q9ERE4, Q9H4A6
Gene Alias 4733401N08Rik; 5730410D03Rik; AW413496; coat protein GPP34; coat-protein; Gmx33; golgi peripheral membrane protein 1, 34 kDa; Golgi phosphoprotein 3; golgi phosphoprotein 3 (coat-protein); golgi protein; golgi-associated protein; GOLPH3; GOPP1; GPP34; MIDAS; Mitochondrial DNA absence factor; trans-Golgi protein GMx33; Vps74
Gene Symbols Golph3
Host Species Rabbit
Immunogen A 14 amino acid peptide from near the amino terminus of human GOLPH3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64083, 66629, 78961
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.