missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLPH3 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67777-1-IG-20UL
This item is not returnable.
View return policy
Description
The GOLPH3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GOLPH3. This antibody recognizes human, mouse, rat antigen. The GOLPH3 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| GOLPH3 | |
| Monoclonal | |
| 1000 μg/mL | |
| FLJ90675; GOPP1; GPP34; MIDAS | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 34 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 3H2A5 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 64083 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction