missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GOLM1 Monoclonal specifically detects GOLM1 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | GOLM1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 5 |
| Conjugate | Alexa Fluor 532 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human GOLM1 (Q8NBJ4-1; Val 40-Leu 401). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?