missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GOLIM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115311
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GOLPH4.
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi-resident protein. It may process proteins synthesized in the rough endoplasmic reticulum and assist in the transport of protein cargo through the Golgi apparatus.
Specifications
| GOLIM4 | |
| Polyclonal | |
| Unconjugated | |
| GOLIM4 | |
| 130 kDa golgi-localized phosphoprotein; 3110027H23Rik; cis Golgi-localized calcium-binding protein; decapacitation factor 10; Df10; GIMPc; Golgi integral membrane protein 4; Golgi integral membrane protein, cis; golgi phosphoprotein 4; Golgi phosphoprotein of 130 kDa; golgi-localized phosphoprotein of 130 kDa; GOLIM4; GOLPH4; GPP130; LOW QUALITY PROTEIN: Golgi integral membrane protein 4; P138; type II Golgi membrane protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27333, 310526, 73124 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O00461, Q5BJK8, Q8BXA1 | |
| GOLIM4 | |
| A synthesized peptide derived from human GOLIM4(Accession O00461), corresponding to amino acid residues E528-E578. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction