missing translation for 'onlineSavingsMsg'
Learn More

Golidocitinib, MedChemExpress

Product Code. 16494108
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494108 10 mg 10mg
16460467 50 mg 50mg
16416817 5 mg 5mg
16415086 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494108 Supplier Medchem Express Supplier No. HY107361
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Golidocitinib (AZD4205) is a selective JAK1 inhibitor, with an IC50 of 73 nM, weakly inhibits JAK2 (IC50>14.7 μM), and shows little inhibition on JAK3 (IC50>30 μM).

Golidocitinib (AZD4205) is a selective JAK1 inhibitor, with an IC50 of 73 nM, weakly inhibits JAK2 (IC50>14.7 μM), and shows little inhibition on JAK3 (IC50>30 μM).

Specifications

Chemical Name or Material Golidocitinib
CAS 2091134-68-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H31N9O2
Quantity 10 mg
Synonym AZD4205
Solubility Information DMSO : 250 mg/mL (510.65 mM; Need ultrasonic)
SMILES C[C@@H](N1CCN(C)CC1)C(NC2=C3C(C(C4=NC(NC5=CN(C)N=C5OC)=NC=C4)=CN3)=CC=C2)=O
Molecular Weight (g/mol) 489.57
Formula Weight 489.57
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.