missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Golgin-97 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114509
This item is not returnable.
View return policy
Description
Positive controls that may be used include: Human Stomach Tissue Lysate.
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome.
Specifications
| Golgin-97 | |
| Polyclonal | |
| Unconjugated | |
| GOLGA1 | |
| 0710001G09Rik; 2210418B03Rik; AW107649; gap junction protein, alpha 4, 37kD; Golga1; golgi autoantigen, golgin subfamily a, 1; Golgi97; golgin A1; golgin subfamily A member 1; golgin-97 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2800, 311919, 76899 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q92805, Q9CW79 | |
| GOLGA1 | |
| GOLGIN97 antibody was raised against a 17 amino acid peptide near the amino terminus of human GOLGIN97. The immunogen is located within amino acids 30 - 80 of GOLGIN97. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction