missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Golgin-97 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530048
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116, mouse brain. Predicted reactivity: Mouse (90%), Rat (90%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome.
Specifications
| Golgin-97 | |
| Polyclonal | |
| Unconjugated | |
| GOLGA1 | |
| 0710001G09Rik; 2210418B03Rik; AW107649; gap junction protein, alpha 4, 37kD; Golga1; golgi autoantigen, golgin subfamily a, 1; Golgi97; golgin A1; golgin subfamily A member 1; golgin-97 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100751002, 2800, 76899 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative; pH 7 | |
| Q92805, Q9CW79 | |
| GOLGA1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Golgin 97. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Hamster, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction