missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Golgin 97 Antibody (S07-9C6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19958-100ul
This item is not returnable.
View return policy
Description
Golgin 97 Monoclonal antibody specifically detects Golgin 97 in Human samples. It is validated for Western Blot
Specifications
| Golgin 97 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S07-9C6 | |
| Unconjugated | |
| gap junction protein, alpha 4, 37kD, golgi autoantigen, golgin subfamily a, 1, golgin A1, Golgin subfamily A member 1, Golgin-97, MGC33154 | |
| A synthetic peptide of human Golgin 97 (Uniprot # Q92805) | |
| 100 μg | |
| Golgi Apparatus Markers | |
| 2800 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction