missing translation for 'onlineSavingsMsg'
Learn More
Learn More
golgi transport 1 homolog B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38088-25ul
This item is not returnable.
View return policy
Description
golgi transport 1 homolog B Polyclonal specifically detects golgi transport 1 homolog B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| golgi transport 1 homolog B | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q9Y3E0 | |
| GOLT1B | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RRVPVLGSLLNLPGIRSFVDKVGESN | |
| 25 μL | |
| Signal Transduction | |
| 51026 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CGI-141, Golgi transport 1 homolog B, golgi transport 1 homolog B (S. cerevisiae), golgi transport 1B, GOT1, GOT1A, hGOT1a, Putative NF-kappa-B-activating protein 470, vesicle transport protein GOT1B, YMR292W | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction