missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Golgi protein 58k Recombinant Rabbit Monoclonal Antibody (23GB5765), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£162.00 - £387.00

Specifications

Antigen Golgi protein 58k
Clone 23GB5765
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30088181
GREENER_CHOICE
View Documents
Invitrogen™
MA552912
100 μL
£387.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30089820
View Documents
Invitrogen™
MA552913
20 μL
£162.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Golgi protein 58k Recombinant Monoclonal antibody specifically detects Golgi protein 58k in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications

Specifications

Golgi protein 58k
Flow Cytometry, Western Blot
Unconjugated
Rabbit
Human
O95954
10841
IgG
Affinity chromatography
Antibody
23GB5765
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1
A synthesized peptide derived from human 58K Golgi protein (50-100AA).
Primary
-20°C
Ftcd
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.