missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Golgi protein 58k Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112254
This item is not returnable.
View return policy
Description
Golgi protein 58k Polyclonal Antibody for Western Blot, IHC (P), IP
Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.
Specifications
| Golgi protein 58k | |
| Polyclonal | |
| Unconjugated | |
| Ftcd | |
| 58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10841 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| O95954 | |
| Ftcd | |
| E. coli-derived Human FTCD fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction