missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Golgi protein 58k Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA19001
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.
Specifications
| Golgi protein 58k | |
| Polyclonal | |
| Unconjugated | |
| Ftcd | |
| 58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10841 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O95954 | |
| Ftcd | |
| Synthetic peptide corresponding to the N Terminus amino acids SQLVECVPNFSEGKNQ (aa 2-18). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction