Learn More
Invitrogen™ Golgi protein 58k Monoclonal Antibody (K92024_1D11), Invitrogen™
Mouse Monoclonal Antibody
£516.00 - £796.00
Specifications
| Antigen | Golgi protein 58k |
|---|---|
| Clone | K92024_1D11 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30200704
|
Invitrogen™
MA555658 |
50 μL |
£516.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30202243
|
Invitrogen™
MA556038 |
200 μL |
£796.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: FSEGKNQEVI DAISGAITQT PGCVLLDVDA GPSTNRTVYT FVGPPECVVE GALNAARVAS RLIDMSRHQG EHPRMGALDV CPFIPVRGVS VDECVLCAQA FGQRLAEELD VPVYLYGEAA RMDSRRTLPA IRAGEYEALP KKLQQADWAP DFGPSSFVPS WGATATGARK FLIAFNINLL GTKEQAHRIA LNLREQGRGK DQPGRLKKVQ GIGWYLDEKN LAQVSTNLLD FEVTALHTVY EETCREAQEL SLPVVGSQLV GLVPLKALLD AAAFYCEKEN LFILEEEQRI RLVVSRLGLD SLCPFSPKER IIEYLVPERG
Golgi protein 58k Monoclonal antibody specifically detects Golgi protein 58k in Human, Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| Golgi protein 58k | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1 | |
| Recombinant human FTCD (immunogen range is 11-330). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Ftcd |
| K92024_1D11 | |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| O88618, O95954 | |
| 10841, 89833 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.