missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Golgi protein 58k Monoclonal Antibody (GT575)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531518
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Pig (85%), Bovine (81%). Positive Control: HepG2, mouse liver. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Golgi Protein, 58K is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool. Mutations in this gene are associated with glutamate formiminotransferase deficiency. Alternatively spliced transcript variants have been found for this gene.
Specifications
| Golgi protein 58k | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O95954, Q91XD4 | |
| Ftcd | |
| Recombinant protein encompassing a sequence within the C-terminus region of human 58K Golgi protein. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| GT575 | |
| Unconjugated | |
| Ftcd | |
| 58 kDa microtubule-binding protein; Formimidoyltetrahydrofolate cyclodeaminase; formimidoyltransferase cyclodeaminase; Formimidoyltransferase-cyclodeaminase; Formiminotetrahydrofolate cyclodeaminase; formiminotransferase cyclodeaminase; formiminotransferase-cyclodeaminase; FTCD; Glutamate formimidoyltransferase; Glutamate formiminotransferase; Glutamate formyltransferase; I79_006831; LCHC1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 10841, 14317 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction